Axygen® Robotic Pipette Tips, Oasis Series, Corning
Fournisseur: Corning
Certificats
A propos de cet article
These tips for Dynamic Device Oasis automation systems are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
- Transparent PP
- Certified RNase-/DNase-free
- Non pyrogenic
The Oasis series robotic tips are engineered to exact dimensions of OEM tips to ensure automation compatibility.
Customers Who Bought This Also Bought