Automation Pipette Tips for Qiagen BioRobot Workstations, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Thermo Scientific provides a solution for your Qiagen BopRobot pipette tips.
These automation pipette tips have been designed, validated, and tested on a Qiagen BioRobot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our BioRobot workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Specifications
- Catalog No:
- 72830-336
- 72830-334
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)