Flask, Media Storage and Dispensing, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This media storage flask is used for sterile dispensing; use with an aseptic filling bell
Bottom 1/2-inch hose connection assures aseptic drainage. With beaded rim. Fabricated from borosilicate glass. Size value represent the value of Stopper size.
Ordering information: Stopper not included.
Customers Who Bought This Also Bought