Cloning Cylinder, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
You can clone a single cell or group of cells by surrounding them with this glass cylinder
Note: 89083-364 and 89083-358 are for use in the Disinfectant Procedure described in the 1992 J.A.O.A.C. Hard Surface Carrier Test for Efficacy Testing of Disinfectants: Collaborative Study. These tubes are glazed on both ends.
Dip the end of the cylinder into a sterile silicone grease before pressing to the bottom of a culture flask to create an isolated well.
Customers Who Bought This Also Bought