Buret, Automatic, 1:5 PTFE Plug, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This simplified design of this automatic buret provides greater strength, resulting in longer service life
It is also easier to repair in the event of breakage, and eliminates hold-up in the immersed tube.
Ordering information: Buret and bottle are available separately, or you can purchase complete item that includes buret, pressure bulb, polyethylene release tube, and reservoir bottle. For replacement stopcock plug, order 89051-922.
Customers Who Bought This Also Bought