Filling Bell Aseptic, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This apparatus is used for aseptic filling of vessels.
- Fabricated from borosilicate glass
Rubber tubing is attached to the top hose connection, the receiving vessel is placed inside the bell.
Customers Who Bought This Also Bought