Tissue Homogenizer, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This highly efficient, all glass, tapered tissue grinder/homogenizer combines two grinding surfaces on a single pestle and tube
The conical area is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1–0.15 mm clearance. With pouring lip. The 10 mm rod on the three larger size pestles may be used with the 89062-242 "Flex-Grip" chuck. The 6 mm rod on the small size is for use with the 89062-238 chuck. Capacities represent volumes with pestle inserted. Fabricated from borosilicate glass.
Ordering information: Tube and pestle may be purchased separately, or you can get the complete item that includes both.
Frequently Bought Together