Tissue Homogenizer, Tenbroeck, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
These all-glass Tenbroeck tissue grinders come with precision-made, interchangeable pestles and tubes
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
The hollow handle permits packing with ice. This unit also features an expanded reservoir and a pouring lip. Designed for tissues such as liver, intestines, and heart. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought