Tissue Homogenizer, Potter-Elvehjem, Micro, Ace Glass Incorporated
Catalog # 71000-520
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacity:0.1 ml
- Description:Potter-Elvehjem Micro Tissue Homogenizer
- Cat. no.:71000-520
- O.D.×H:4 x 65 mm
- Overall Length:110 mm
- Supplier No.:8357-03
Specifications
About this item
This micro size tissue homogenizer is for extremely precise work
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
This unit is designed for delicate hand operation. A reservoir and pouring lip are incorporated into the design. Fabricated from borosilicate glass.
Frequently Bought Together