Tissue Homogenizer, Potter-Elehjem, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
When using this Potter-Elehjem tissue homogenizer, homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
The mortar is not ground, and the pestle contact surface is PTFE, threaded onto a stainless steel shaft. Tube fabricated from borosilicate glass.
Customers Who Bought This Also Bought