Conductivity/TDS Standard, 20 mS/cm (20000 µmho/cm) at 25 °C, 11000 ppm as NaCl, Ricca Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.