Aluminum Standards for AA Spectroscopy, Ricca Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
Caution: 10,000 ppm Al = 10,000 mg/l Al = 10 mg/ml Al= 10,000 µg/ml Al
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.