PolarSafe™ Stainless Steel Specialty Freezer Racks, Argos Technologies
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Argos PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel
Argos PolarSafe™ Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright and Chest ultra low freezers.
Handles at both ends accommodate freezers with right or left handed doors.
Ordering information: Additional freezer racks available. Contact your Avantor sales representative for details.
Customers Who Bought This Also Bought