Nalgene® Filter Units, 115 ml, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Complete filter units.
- Choose from CN, aPES, PES, or SFCA membranes
- Use 0.2 µm pore size for sterile filtration and 0.45 µm pore size for clarification and particle removal
Note: For research use only. Not for in vitro diagnosis or parenterals.
Presterilized, individually packaged one-piece vacuum filter units with 115 ml capacity are ideal for easy, economical filtration of 50 to 115 ml of aqueous fluids. Upper and lower chambers with graduations do not separate; simply remove vacuum hose after filtration and pour filtered media out. Catalog number, membrane, pore size, lot number and expiration date are printed on upper chamber of each filter unit.
Specifications
- Catalog No:
- 28199-036
- 28199-048
- 28199-032
- 28199-034
- 28199-030
- 73520-980
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)