Ampule Snapper, Disposable, WHEATON®, DWK Life Sciences
# de catalogue 66009-125
Fournisseur: DWK Life Sciences
Small Business Enterprise
Spécifications
- Description:Ampule Snapper
- For:1 - 2 ml ampules
- Supplier No.:177105
Spécifications
A propos de cet article
Disposable ampule snapper protects the user against glass splinters and specially designed for 1 to 2 ml samples.
It prevents cross contamination between samples.
Customers Who Bought This Also Bought