Dropping Bottle, with Tip and Cap, LDPE, WHEATON®, DWK Life Sciences
Supplier: Avantor
Certificates
About this item
WHEATON Dropping Bottles are made of LDPE (Low Density Polyethyleye).
Dropping bottles with tips can be used to dispense a stream of liquid or a non-controlled drop. Dropping bottles, tips and caps are packaged separately and bottles are autoclavable.
Customers Who Bought This Also Bought