Dispensing Bottle, LDPE, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
The WHEATON Dispensing Bottle is made from low denisty polyethylene.
The dispensing bottle is an excellent option for mild and strong buffers, and offers good chemical resistance. To dispense the contents, the spout is raised and to seal the bottle the spout is depressed.
It is translucent in color and made of very flexible but high impact strength LDPE.
Customers Who Bought This Also Bought