À propos de cet article
The Barnstead Pacific RO water purification systems provides up to 240L/day of Type 3 (Reverse Osmosis) water from tap water
The Pacific RO systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. The 30L and 60L reservoirs may also be mounted on the wall, on the bench, or under the bench.
The Pacific RO systems are ideal for producing type 3 (RO) for the rinsing of glassware, supplying lab equipment such as autoclaves, chambers, or water baths, or the preparation and diluting of buffers, reagents and media.
All Pacific RO systems include: quick discconnects to make changing consumables easy, and all core consumables and tubing. Pacific RO systems are offered in two output volumes, 7L/hr and 20L/hr. External reservoirs are required and are offered in three different sizes, 30L, 60L or 100L.
Spécifications
- Catalog No:
- 89230-438
- 89230-442
- W×D×H:
- 372×330×603 mm (14.6×13×23.7")
- 372×330×603 mm (14.6×13×23.7")
Spécifications
Informations connexes
In addition, VWR CATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWR[email protected] for further details.