About this item
The Barnstead LabTower EDI water purification systems provides up to 200L/day of Type 1, ultrapure (18.2 megohm) water or Type 2 (10-15 megohm) water from tap water
The LabTower EDI systems are completely free-standing units which require no bench or wall space. The unit comes standard with casters for easy moving.
The LabTower EDI systems are suited to meet a wide range of applications needs. Type 1 water can be used for cell and tissue culture, PCR, DNA sequencing, electrophoresis, HPLP, GC-MS or ICP-MS for example. Type 2 water can be used for rinsing glassware, preparing and diluting buffers, reagents or media for example.
All LabTower EDI systems include: quick discconnects to make changing consumables easy, UV lamp, built in 100L reservoir and all core consumables and tubing. LabTower EDI systems are offered in two output volumes, 15L/hr or 30L/hr. Both models come standard with a 100L integrated reservoir.
Specifications
- Catalog No:
- 89230-450
- 89230-452
- Feed water:
- Softened water
- Softened water
- Production flow rate:
- 15 L/h
- 30 L/h
- W×D×H:
- 450×580×1500 mm (17.7 × 22.8 × 59.1")
- 450×580×1500 mm (17.7 × 22.8 × 59.1")