Label "Infant Drowning Warning", National Marker
# de catalogue 94004-308
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:Warning labels
- Material:PS Vinyl
- Cat. no.:94004-308
- Legend:Infant Drowning Label (Bilingual)
- Dimensions:127×76.2 mm (5×3")
- Supplier No.:BID1
Spécifications
A propos de cet article
Warns of infant drowning hazard in 4 to 6 gallon plastic and metal buckets.
Customers Who Bought This Also Bought