NMC Glow Lockout Tag
# de catalogue 94014-978
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Material:Glow Unrip Vinyl
- Size:15.24 x 7.62 cm (6 x 3")
- Cat. no.:94014-978
- Legend:Danger This Tag and Lock To Be Removed..
- Supplier No.:GLTAG1
Spécifications
A propos de cet article
Prevent accidental or unexpected start up or release of stored energy when working on equipment and machiner15.24cm x 7.62cm (6" x 3").
Customers Who Bought This Also Bought