Overlaminate Tags, Blank, National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Personalize your tags with specific information and instructions, peel away liner and seal overlamiante onto tag to protect message.
Unrippable vinyl with clear polyester self-laminating cover.
Customers Who Bought This Also Bought