Zoology/Ornithology Cabinet
# de catalogue 470022-340
Fournisseur: Lane Science
Spécifications
- Description:Zoology/ornithology cabinet
- Cat. no.:470022-340
- Supplier No.:201
Spécifications
A propos de cet article
With ample space for up to eighteen trays and a pestproof, airtight seal, this cabinet provides optimal protection for bird and mammal study skins.
Size: 46½×28×42"
Weight: 246 lbs.
Ordering information: The trays are sold separately.
Caution: Note: item is shipped motor freight.
Customers Who Bought This Also Bought