Ward's® Zebra Mussel
# de catalogue 470001-338
Fournisseur: Avantor
Spécifications
- Size:1/4–1/2"
- Cat. no.:470001-338
- Packaging:Jar of 10
- Supplier No.:WARD470001-338
Spécifications
A propos de cet article
Formalin Preserved
This exotic species was introduced to the United States’ Great Lakes area and is carried in the ballast water of ships entering the Saint Lawrence Seaway. Their prolific growth causes problems in municipal water systems.
Customers Who Bought This Also Bought