Your Body Microslide
Fournisseur: SUPERTEK SCIENTIFIC LLC
Certificats
A propos de cet article
Illustrates body tissues and organs
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Small intestine (50X), Bronchial tree, Heart (cs, 6X), Bone (cs, 150X), Muscle (cs, 900X), Neuron (cs, 400X), Pancreas gland (190X), Skin (cs, 80X), and White blood cells (em, 25,000X).
Customers Who Bought This Also Bought