Ward's® Prepared Yeast, Genetic (YEAD) Agar
Fournisseur: Culture Media & Supplies
Certificats
A propos de cet article
For Maintaining Haploid Yeast Cultures
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Yeast extract-adenine-dextrose growth media.
Customers Who Bought This Also Bought