Ward's® Prepared Yeast, Genetic (MIN) Agar
# de catalogue 470180-814
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Packaging:Plates
- Description:Yeast Genetic (MIN) Agar
- Cat. no.:470180-814
- Supplier No.:WARD470180-814
Spécifications
A propos de cet article
For Demonstration of Typtophan Dependance or Independence in Yeast
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Ordering information: Some of these products contain living or perishable materials
Delivery: Prepared plates will ship via overnight or 2nd day delivery.
Prepared bottles and tubes do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of freight. During checkout, please specify a delivery date for your specimens to arrive.
Prepared bottles and tubes do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of freight. During checkout, please specify a delivery date for your specimens to arrive.
Customers Who Bought This Also Bought