Wooden Meter and Half-Meter Sticks
# de catalogue 470005-628
Fournisseur: American Scientific
Small Business Enterprise
Spécifications
- Description:Metric/imperial ruler
- Material:Wood with metal ends
- Cat. no.:470005-628
- Length:1 m (3.2 ft.)
- Supplier No.:2192-98
Spécifications
A propos de cet article
Good quality hardwood meter and half-meter sticks with a natural finish.
- Permanently engraved scales
- One side is graduated in cm with 1mm divisions
- Other side is graduated in inches with 1/8-inch divisions
Meter sticks are 6mm thick x 25.4mm wide.
Customers Who Bought This Also Bought