Wooden Crystal Models
# de catalogue 470154-308
Fournisseur: DAS Woodworking
Spécifications
- Description:Wooden Crystal Model Set/12
- Cat. no.:470154-308
- Supplier No.:DEFAULT VPN
Spécifications
A propos de cet article
Colorful additions to your mineraology classroom, these carefully crafted models are painted with durable enamel in colors that simulate the actual crystal.
Measuring from 3 to 6" in length, the models are precisely shaped with correct angles to illustrate the six basic crystal structures: isometric, tetragonal, hexagonal, orthorhombic, monoclinic, and triclinic. Each is color-coded for clear crystal representations. Includes a teacher’s guide and all models in a compartmented storage box. Set of 12.
Customers Who Bought This Also Bought