Wood Chalkboard Compass
# de catalogue 470014-878
Fournisseur: Aakron Rule
Spécifications
- Description:Wood chalkboard compass
- Cat. no.:470014-878
- Supplier No.:100014
Spécifications
A propos de cet article
This wooden compass draws symmetrical circles every time. A rubber tip prevents slipping to help you maintain the center point.
Size: 17"
Circle diameter: up to 50"
Customers Who Bought This Also Bought