Wire Leads with Alligator Clips
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Easily connect electrical devices that are lacking banana plugs.
These wire leads are extra flexible and are plastic coated with alligator clips at both ends. The leads are 56 cm long and the alligator clips feature a protective vinyl sheath.
- Flexible plastic coated wire
- 18 gauge wire
These wire leads are extra flexible and are plastic coated with alligator clips at both ends. The leads are 56 cm long and the alligator clips feature a protective vinyl sheath.
Customers Who Bought This Also Bought