Wire Leads with Alligator Clips
Fournisseur: AMERICAN EDUCATIONAL PROD
Certificats
A propos de cet article
These wire leads are made of high quality, flexible, 18 gauge, Hypalon coated wire and feature insulated alligator clips at both ends.
Each lead measures 90 cm.
Customers Who Bought This Also Bought