Windup Fiberglass Measuring Tapes
# de catalogue 470110-722
Fournisseur: SI Manufacturing
Spécifications
- Length:10 m (33 ft.)
- Description:Fiberglass wind-up tape with plastic case, metric
- Cat. no.:470110-722
- Supplier No.:45200
Spécifications
A propos de cet article
Calibrated in both inches and centimeters, these fiberglass tapes will find use in indoor and outdoor measurements activities.
The 5' version is self-retracting, others are wind-up.
Customers Who Bought This Also Bought