Windsocks, HUFFYS AIRPORT WINDSOCKS
# de catalogue 470106-706
Fournisseur: HUFFYS AIRPORT WINDSOCKS
Spécifications
- Description:Windsock, 4" x 15", Orange, GLOBE Approved Product
- Cat. no.:470106-706
- Supplier No.:2X15O
Spécifications
A propos de cet article
A great addition to your outdoor weather station, these airport quality windsocks are bright orange with reinforced eyelets. Brackets are attached to the windsock by quick lock nylon binding strips so they are easy to replace. Size of windsock: 4" x 15". Size of bracket: 4".
Customers Who Bought This Also Bought