Wind Generator
# de catalogue 470003-064
Fournisseur: VWR International
Spécifications
- Description:Wind Generator
- Cat. no.:470003-064
- Supplier No.:WLS1755-57
Spécifications
A propos de cet article
A small scale demonstration of renewable energy, this wind generator can be used with a voltmeter or datalogger to measure and monitor output.
- Size: 15.2×10.2×15.2 cm
It can also be used to illuminate an LED and illustrate the conversion of energy. Its low-inertia electric motor functions as a generator and is driven by a plastic impeller.
A cold air blower can be used to turn the impeller resulting in a low voltage DC output at the two 4 mm sockets.
Customers Who Bought This Also Bought