Butterflies Of The World Poster
# de catalogue 470015-142
Fournisseur: FEENIXX PUBLISHING, INC.
Spécifications
- Length:92 cm
- Width:61 cm
- Description:Butterflies Poster
- Cat. no.:470015-142
- Finish:Unlaminated
- Supplier No.:A221
Spécifications
A propos de cet article
Colorful Butterfly Illustrations
- These beautiful insects are grouped by continent or other geographic area
This popular poster shows over 100 of the most colorful butterflies, as well as a few moths. The captions cite common name, biological name, and where they are found.
Customers Who Bought This Also Bought