Wide-Mouth Polypropylene Cylinders
Fournisseur: SI Manufacturing
Certificats
A propos de cet article
Chemical and Heat Resistant to 80°C.
- Wide mouth is ideal for performing density tests
- Molded, blue over-printed graduations
- Aligned with GLOBE protocols
The squat design increases stability on the bench while the low form requires less overhead storage space.
Spécifications
- Catalog No:
- 470020-850
- 470020-848
- 470150-800
- 470150-424
- 470020-852
- Form:
- Tall
- Tall
- Tall
- Tall