Whose Fingerprints Were Left Behind Lab Activity
# de catalogue 470127-994
Fournisseur: Edvotek
Spécifications
- Description:Whose Fingerprints Were Left Behind Kit
- No. of Lab Groups:10
- Cat. no.:470127-994
- Supplier No.:S-91
Spécifications
A propos de cet article
Analyze Identifying Evidence
- Grade Level 10-College
- Links to a real world scenario
After a crime has been committed, the evidence left behind can point to the potential culprit. Fingerprints and blood stains are key pieces of evidence, helping to identify and convict a criminal. In this experiment, students will use hands-on scientific methods to solve a crime by dusting for fingerprints, and use fluorescent dust to search for and identify trace amounts of blood.
Customers Who Bought This Also Bought