White Blood Cell, Human Slide
# de catalogue 470183-018
Fournisseur: Avantor
Spécifications
- Description:White Blood Cell, Human Slide
- Cat. no.:470183-018
- Stain:Giemsa
- Supplier No.:WARD470183-018
- Orientation:Smear
Spécifications
A propos de cet article
Huge component within the immune system
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Demonstrates white blood cells and platelets. Smear (buffy coat). Giemsa.
Customers Who Bought This Also Bought