What Rock Is It? Study Kits
Fournisseur: SCOTT RESOURCES 2, LLC SE
Certificats
A propos de cet article
Test your student's geology skills with this educational and fun activity. Students receive a bag of unlabeled rock specimens, then must test for physical and chemical properties in order to identify each rock sample. The easy-to-follow flowchart aids students in their classification decisions.
Kit includes:
• Samples and study charts for 30 student groups
• Large specimens for easy examination
• Chart with mounted specimens
• Detailed study guide with physical properties and descriptions of more than 15 rocks, minerals, and ores
Customers Who Bought This Also Bought