What do Seababies tell us about Pollution Lab Activity
# de catalogue 470024-486
Fournisseur: EDVOTEK
Spécifications
- Description:What do Seababies tell us about Pollution Lab Activity
- No. of Lab Groups:6
- Cat. no.:470024-486
- Supplier No.:DD-304
Spécifications
A propos de cet article
Investigate Pollution
- Simulates environmental incident
- Open or guided inquiry
Demonstrate how pollutants have deleterious consequences on local seawater ecology. In this experiment, the brine shrimp, Artemia franciscana, are hatched from eggs in artificial seawater and the effect of various simulation toxins are added. The effect of simulated toxicants will be monitored by their impact on growth and survival.
Customers Who Bought This Also Bought