Westminster Motor Kit
# de catalogue 470121-102
Fournisseur: VWR International
Spécifications
- Description:Westminster Motor Kit
- Cat. no.:470121-102
- Supplier No.:PH1239
Spécifications
A propos de cet article
For class model building.
Materials are supplied for building six complete motor models. Includes 12 magnets, six armatures, six pieces of mild steel yokes, six support bases, six support shafts, 24 connecting rivets, 12 split pins, one reel of 25 s.w.g., PVC tinned copper wire, two reels of cellotape, 300mm rubber tubing, and instructions.
- Build 6 model motors
The kit comprises all the part required to build 6 model motors as in the electromagnetic kit. Each kit contains 12 pieces of magnets, 6 pieces of armatures, 6 pieces of mild steel yokes, 6 pieces of support bases, 6 pieces of shaft, 24 pieces of rivets, 12 pieces of split pins, 1 reel of 26 s.w.g. PVC tinned copper wire, 2 reels of cellotape, 300 mm rubber tubing of 3 mm bore.
Customers Who Bought This Also Bought