Weather Vane, HAND2MIND INC.
# de catalogue 470012-338
Fournisseur: HAND2MIND INC. MO
Spécifications
- Description:Weather Vane
- Cat. no.:470012-338
- Supplier No.:8-875
Spécifications
A propos de cet article
Ideal for Simple Wind Speed Experiments
- Can be permanently mounted or handheld
Wind direction and velocity are clearly communicated with this practical instrument. Arrows indicating North, South, East, and West are clearly marked. This simple weather vane can be permanently mounted or used as a hand held tool.
Customers Who Bought This Also Bought