ContactUsLinkComponent
Wave Velocity Apparatus
Wave Velocity Apparatus
# de catalogue 470039-916
Fournisseur:  VWR International
APPARATUS WAVE VELOCITY
Wave Velocity Apparatus
# de catalogue 470039-916
Fournisseur:  VWR International
Spécifications
  • Description:
    Wave Velocity Apparatus
  • Input voltage:
    110V
  • Cat. no.:
    470039-916
  • Supplier No.:
    PH0770
Spécifications
A propos de cet article
This self-contained apparatus explores wave velocity.
  • Function Generator Included
  • Easy-To-Adjust Wavelength
  • Apparatus Explores Wave Velocity
Teachers use it to demonstrate the live wave motion that is generated using linear rope and thread. With the provided function generator, the waves generate nodes and antinodes, and their full wave length or half wave length are measurable.
The wavelength can be changed with the help of variation in frequency and stress generated with different weights on the one end of rope and thread.
Customers Who Bought This Also Bought