Water Still
# de catalogue 470003-602
Fournisseur: Dynalon
Spécifications
- Flow rate:4 L/hr.
- Description:Water Still, 230 V
- Cat. no.:470003-602
- Supplier No.:4000-0000
- Width:6"
Spécifications
A propos de cet article
Two capacities available; 2.4L/hr.and 4.0L/hr.
- Complies to ASTM and ISO standards
- Pyrogen free single distilled water
- Easy to clean – no glass dismantling
- Bulit-in condenser to cool water
Still features a thermostat that protects it in the event of a water supply failure and automatically resets when the still cools. It’s easy to clean by adding cleaning agents through a built-in funnel. It also comes with a drainage stopcock and metal stand for mounting to a wall or tabletop.
Ordering information: Available to U.S. customers only.
Customers Who Bought This Also Bought