Washers
Catalog # 470019-392
Supplier: Avantor
Specifications
- Description:Washers
- Cat. no.:470019-392
- Supplier No.:WARD470019-392
Specifications
About this item
These small items are commonly used as weights. Package of twelve.
- Flat steel 2P
- Size: 1" O.D. x 7/16" I.D
- weight: approximately 6 g each
Customers Who Bought This Also Bought