A propos de cet article
Need all your services in one place or looking to add a wall of cabinets that include services? The Wall Service Bench offers storage and services all in one unit.
- Constructed of solid oak and oak veneer plywood
- Outer ends of the unit offer a full height locking doors and one adjustable shelf
The Wall Service Bench features your choice of a 1" solid epoxy resin or phenolic resin top with a 18W×15D×11"H epoxy resin sink and two combination cold water/gas fixtures. The sink area allows access to the plumbing and provides a large storage space with two GFI protected AC duplex electrical outlets. There is enough room in the sink area to allow two 24" D tables to be butted against the unit for use as a peninsula work area. The outer ends of the unit offer four storage solutions: doors with adjustable shelves, doors/drawer combination with one adjustable shelf, four drawers/doors with one adjustable shelf and four drawers. All drawers and doors have locks and ADA compliant pulls. The unit is finished with a chemical resistant, earth-friendly UV finish and includes rubber base molding.
Sink dimensions: 18L×15W×11"D
Spécifications
- Catalog No:
- 470117-968
- 470117-964
- 470117-950
- 470117-936
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green