ContactUsLinkComponent
Wall Mounted Eyewash Station
Wall Mounted Eyewash Station
# de catalogue 470205-492
Fournisseur:  WaterSaver Faucet
WALL MOUNT EYEWASH STATION
Wall Mounted Eyewash Station
# de catalogue 470205-492
Fournisseur:  WaterSaver Faucet
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Wall Mounted Eyewash
  • Cat. no.:
    470205-492
  • Supplier No.:
    EW814
Spécifications
A propos de cet article
This wall-mounted unit quickly and gently rinses contaminates away from both eyes simultaneously.
  • Stainless steel bowl for safe, simple use
  • Two heads deliver flood of water for hands-free eye rinsing
  • Heavy-duty pipe and fittings resist corrosion
Featuring two GS-Plus™ (Gentle Spray) Heads, this low-density polyethylene filter removes scale, deposits, and debris from the water and keeps water line impurities out of users' eyes.
A push handle activates the eye wash, which allows for hands free operation.
Certifications: ANSI-complaint identification sign included.
Customers Who Bought This Also Bought