Single-Scale Precision Cylinder
# de catalogue 470148-742
Fournisseur: VWR International
Spécifications
- Capacity:10 ml
- Graduation Interval:0.1 ml
- Graduation range:0–10 ml
- Form:Tall
- Cat. no.:470148-742
- Tolerance:0.2 ml
- Supplier No.:V3022-10
Spécifications
A propos de cet article
Our high-quality borosilicate cylinders meet ASTM standards.
- Cylinders Calibrated to Contain
- Borosilicate Cylinders
- Uniform Wall Thickness
- Molded Base
- Graduated Metric Scale
Cylinders feature uniform wall thickness, white enamel, a single and graduated metric scale, pour-out, and a molded base.
They can be used with acids, salt solutions, and organic solvents.
Customers Who Bought This Also Bought