VWR® Lab Notebook
# de catalogue 470151-176
Fournisseur: Heathrow Scientific
Spécifications
- Ruling:Grid
- Description:VWR® lab notebook
- Cat. no.:470151-176
- Supplier No.:HEA8610AV
Spécifications
A propos de cet article
Keep track of lab data and results with this convenient lab notebook.
- Available in grid versions
- Hardcover
- 100 pages
- Size: 8.58" x 11.06"
- Includes numbered pages, “continued from” and a table of contents
In addition to blank pages for student records and remarks, it includes basic instructions on how to properly do a lab and state the outcomes. A full page periodic chart of the elements and conversion charts are also included for easy access to reference information.
Customers Who Bought This Also Bought